Learn More
Alkali Scientific SmartBlue Lab Safety Glasses
SDP
Catalog No. 502389168
Encompass_Preferred
SmartBlue Plus Viewing Glasses offer exceptional protection for researchers working with bright blue LED and UV light illuminators. The polycarbonate filter lenses are shatterproof, scratch-resistant, and block light in the UV (280nm) to blue (490nm) wavelengths. These glasses are ideal for general protection when working with hazardous wavelengths and for precision DNA band cutting on a UV or blue light transilluminator. While blocking hazardous UV and blue light, the glasses allow clear visualization of DNA bands stained with Ethidium Bromide, SmartGlow™, SYBR Green, and other fluorescent dyes.
Catalog No. 50-238-9168 Supplier Alkali Scientific Supplier No. RE4000VG
SmartBlue Plus Viewing Glasses offer exceptional protection for researchers working with bright blue LED and UV light illuminators. The polycarbonate filter lenses are shatterproof, scratch-resistant, and block light in the UV (280nm) to blue (490nm) wavelengths. These glasses are ideal for general protection when working with hazardous wavelengths and for precision DNA band cutting on a UV or blue light transilluminator. While blocking hazardous UV and blue light, the glasses allow clear visualization of DNA bands stained with Ethidium Bromide, SmartGlow™, SYBR Green, and other fluorescent dyes.