Learn More
Research Products International Corp SmartBoats, White, 9.5 x 9.5 x 2.5 cm, 250 per Case
SDP
Catalog No. 501367505
Encompass
Simple and cost effective solution to reduce plastic waste. Grease proof boats withstand temperature up to 210° C. Easy to handle SmartBoats are naturally biodegradable and FSC Certified.
Catalog No. 50-136-7505 Supplier Research Products International Corp Supplier No. 142162
Simple and cost effective solution to reduce plastic waste. Grease proof boats withstand temperature up to 210° C. Easy to handle SmartBoats are naturally biodegradable and FSC Certified.