Learn More
Braintree Scientific SMARTBOX Programmable Lab Control Unit - Four Gas Output
SDP
Supplier:  Braintree Scientific EA34000
Encompass
SMARTBOX™ Lab Control Unit Compliant with AVMA Guidelines Operates at a fixed flow rate based on the external CO2 source. The Controller automates on/off gas flow at the push of a button, delivering CO2 to individual cage or Euthanex lid. The EA-34000 series has four gas outputs and four preset times that are programmable. This unit is designed for small scale use by individual investigators in their laboratories. Includes Lab Control Unit and four EA-1130 lab unit hose. Lids and chambers not included! Lab Control Unit Features: Treats animals at individual cage level Designed for investigators that need to retrieve animals quickly Utilizing the full range of Euthanex Cage Lids or existing induction chamber Minimizes operator error and excessive gas use Automatic shut-off saves gas and protects personnel Double throw switches allow for two different time settings for each output Easily attaches to wall **Sold for research purposes only and not for human use**
Catalog No. 50-195-4735
 Restricted Item