Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LevGo smartBoxHolder™ Universal Dispensing Box Holder

Catalog No. 15555001
Change view
Click to view available options
No. of Compartments:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Compartments
15-555-001 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-555-001 Supplier LevGo Supplier No. 50101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sturdy ABS plastic holder provides maximum usability and versatility in any area with minimal storage space. Holder easily expands from 3.75 inches to 6 inches for a wide variety of dispensing boxes, containers, and small electronics.

  • Patent pending design provides maximum usability and versatility in the lab
  • Constructed of sturdy ABS plastic to accomodate a variety of laboratory equipment
  • Easily expands from 3.75 inches to 6 inches (95 mm to 155 mm)
  • Indexed mounting system allows for 180° rotation in 45° increments
  • Built-in port for power cords to pass through
  • Can be mounted on most vertical surfaces

Specifications

Product Type Glove Box Holder
Color Clear
Width (English) 3.75 to 6 in.
Width (Metric) 95 to 155 mm
Holds Disposable Glove Boxes
Material ABS
Mounting Wall Mount
No. of Compartments 1
Capacity 1 Box
Includes Double-sided tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.