Learn More
Wanzl SmartCell™ Chrome Shelving Divider
Description
- Allow for custom compartmentalization of the SmartCell™ basket system.
- Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval
Specifications
Specifications
| Color | Chrome |
| Material | Steel |
| Dimensions (L x W) | 12 x 6 in. |
| For Use With (Application) | Shelving |
| For Use With (Equipment) | SmartCell™ |
| Includes | 1 Divider |
| Width (English) | 6 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.