Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl SmartCell™ Shelving Pedestal Frame Hang Bar

Catalog No. 11222176
Change view
Click to view available options
Width (English):
18 in.
36 in.
48 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-222-176 48 in.
11-222-174 18 in.
11-222-175 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-222-176 Supplier Wanzl Supplier No. T.H48HB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pedestal Frame Hang Bar for SmartCell™ Shelving.

  • Pedestal Frame Hang Bar
  • Replacement part for SmartCell™ Shelving

Specifications

For Use With (Application) Shelving
Includes 1 Hang Bar
Width (English) 48 in.
With Casters No
Color Chrome
For Use With (Equipment) SmartCell™
Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.