Learn More
Takara Bio SMARTer PCR cDNA Synthesis Kit, 20 rxns
SDP
Catalog No. NC1948275
Encompass
A kit for performing twenty cDNA syntheses from nanogram amounts of total or poly A+ RNA. The kit utilizes our SMART (Switching Mechanism At 5' end of RNA Template) technology coupled with PCR amplification to generate high yields of full-length double-stranded cDNA suitable for various applications, including Clontech PCR-Select cDNA subtraction, nondirectional cloning, and preparation of complex cDNA probes. The SMARTer PCR cDNA Synthesis Kit includes the same components as the original SMART PCR cDNA Synthesis Kit plus an improved SMARTer II A oligo and SMARTScribe Reverse Transcriptase, which provide higher specificity and increased yield.
Catalog No. NC1948275 Supplier Takara Bio Supplier No. 634926
A kit for performing twenty cDNA syntheses from nanogram amounts of total or poly A+ RNA. The kit utilizes our SMART (Switching Mechanism At 5' end of RNA Template) technology coupled with PCR amplification to generate high yields of full-length double-stranded cDNA suitable for various applications, including Clontech PCR-Select cDNA subtraction, nondirectional cloning, and preparation of complex cDNA probes. The SMARTer PCR cDNA Synthesis Kit includes the same components as the original SMART PCR cDNA Synthesis Kit plus an improved SMARTer II A oligo and SMARTScribe Reverse Transcriptase, which provide higher specificity and increased yield.