Learn More
CLICKBIO INC Smarter Reagent Fill Station Tubing Set (10ft.)
Catalog No. NC2885875
Encompass
The Tubing Set consists of a fill tube, a drain tube, and clips to attach to the rail on the Deck Module. Case of 10 x 10ft tubing sets.
Catalog No. NC2885875 Supplier CLICKBIO INC Supplier No. CBAFSTSD10
The Tubing Set consists of a fill tube, a drain tube, and clips to attach to the rail on the Deck Module. Case of 10 x 10ft tubing sets.