Learn More
CLICKBIO INC Smarter Reagent Fill Station
Catalog No. NC2279849
Encompass
ClickBio’s Smarter Reagent Fill Station is an on deck reservoir that continuously replenishes itself from an off deck reservoir. This allows a single deck position to supply an unlimited volume of reagent. SBS footprint. Packaged with a Smarter Reagent Fill Station Tubing Set.
This item is not returnable. View return policy
Catalog No. NC2279849 Supplier CLICKBIO INC Supplier No. CBAFS1
Add to Cart
Add to Cart
Request a Quote
ClickBio’s Smarter Reagent Fill Station is an on deck reservoir that continuously replenishes itself from an off deck reservoir. This allows a single deck position to supply an unlimited volume of reagent. SBS footprint. Packaged with a Smarter Reagent Fill Station Tubing Set.