Learn More
MilliporeSigma™ SmartFlare™ NODAL, Human, Cyanine3 RNA Detection Probe
Catalog No. SF483MI Shop All MilliporeSigma Products
Change view
Click to view available options
NODAL, Human, Cyanine3 RNA Detection Probe
Catalog No.
SF483MI NODAL, Human, Cyanine3 RNA Detection Probe
1 options
Catalog No. SF483MI Supplier MilliporeSigma™ Supplier No. SF483
Used as biomarker to detect sequence specfic RNA.
  • Fluorescent reagent, enables sequence specific mRNA
  • MicroRNA expression detection at single cell resolution
  • Allows researchers to identify cells expressing biomarkers of interest
  • Enrich sort cells based on individual expression levels through FACS
  • Compatible with downstream functional analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Includes VEE-OKS-iG RNA, B18R RNA, Human Recombinant B18R Protein, Carrier-Free, Human iPS Reprogramming Boost Supplement II