Learn More
MilliporeSigma™ SmartFlare™ NODAL, Human, Cyanine5 RNA Detection Probe
Catalog No. SF494MI Shop All MilliporeSigma Products
Change view
Click to view available options
NODAL, Human, Cyanine5 RNA Detection Probe
Catalog No.
SF494MI NODAL, Human, Cyanine5 RNA Detection Probe
1 options
Catalog No. SF494MI Supplier MilliporeSigma™ Supplier No. SF494
Used as biomarker to detect sequence specfic RNA.
  • Fluorescent reagent, enables sequence specific mRNA
  • MicroRNA expression detection at single cell resolution
  • Allows researchers to identify cells expressing biomarkers of interest
  • Enrich sort cells based on individual expression levels through FACS
  • Compatible with downstream functional analysis
TRUSTED_SUSTAINABILITY
Specifications
No. of Reactions 252 Reactions/Kit