Learn More
Braintree Scientific SmartGlow, Loading Dye with Safe Green Stain, 1.0ml
SDP
Catalog No. 501954722
Encompass
SmartGlow™ Nucleic Acid Stain Ideal for excitation of nucleic acids with UV of blue light. Unlike EtBR, SmartGlow is non mutagenic and does not require special methods of disposal. Replaces hazardous ethidium bromide (EtBr) Better sensitivity than EtBr (down to 0.1ng) Excitation of both UV and blue light Ships at ambient temperature (store at 4C) Two types: PS (Pre-Stain) or LD (Loading Dye) Compatible with SmartBlue™ **Sold for research purposes only and not for human use**
Catalog No. 50-195-4722 Supplier Braintree Scientific Supplier No. E4500LD
SmartGlow™ Nucleic Acid Stain Ideal for excitation of nucleic acids with UV of blue light. Unlike EtBR, SmartGlow is non mutagenic and does not require special methods of disposal. Replaces hazardous ethidium bromide (EtBr) Better sensitivity than EtBr (down to 0.1ng) Excitation of both UV and blue light Ships at ambient temperature (store at 4C) Two types: PS (Pre-Stain) or LD (Loading Dye) Compatible with SmartBlue™ **Sold for research purposes only and not for human use**