Learn More
Alkali Scientific SmartGlow™ Safe Green Pre Nucleic Acid Stain, Loading Dye, 1mL
SDP
Catalog No. 503027487
Encompass_Preferred
SmartGlow Nucleic Acid Stain is ideal for excitation of nucleic acids with UV or visible blue light transilluminators. Unlike EtBR, SmartGlow it is non-mutagenic and does not require special methods of disposal. SmartGlow PS (Pre Stain) can be used as a direct replacement for Ethidium Bromide in agarose and polyacrylamide gel electrophoresis. The stain emits green fluorescence when bound to dsDNA, ssDNA and RNA. SmartGlow PS exhibits excitation peaks at 290nm and 490nm, allowing it to be used with UV and blue light. SmartGlow LD (Loading Dye) is supplied at 6X concentration and is added directly to the samples. No dye needs to be added to the gel or running buffer. After electrophoretic separation, view and document your results using a UV or blue light illuminator. SmartGlow LD emits green fluorescence when bound to dsDNA, ssDNA and RNA. Replaces hazardous EtBr Excitation by UV or blue light Non-mutagenic, environmentally safe Two types: PS (Pre-Stain) LD (Loading Dye)
Catalog No. 50-302-7487 Supplier Alkali Scientific Supplier No. ES4500LD
SmartGlow Nucleic Acid Stain is ideal for excitation of nucleic acids with UV or visible blue light transilluminators. Unlike EtBR, SmartGlow it is non-mutagenic and does not require special methods of disposal. SmartGlow PS (Pre Stain) can be used as a direct replacement for Ethidium Bromide in agarose and polyacrylamide gel electrophoresis. The stain emits green fluorescence when bound to dsDNA, ssDNA and RNA. SmartGlow PS exhibits excitation peaks at 290nm and 490nm, allowing it to be used with UV and blue light. SmartGlow LD (Loading Dye) is supplied at 6X concentration and is added directly to the samples. No dye needs to be added to the gel or running buffer. After electrophoretic separation, view and document your results using a UV or blue light illuminator. SmartGlow LD emits green fluorescence when bound to dsDNA, ssDNA and RNA. Replaces hazardous EtBr Excitation by UV or blue light Non-mutagenic, environmentally safe Two types: PS (Pre-Stain) LD (Loading Dye)