Learn More
Braintree Scientific SmartGlow, Safe Green Pre Stain, 1.0ml
SDP
Catalog No. 501954723
Encompass
SmartGlow™ Nucleic Acid Stain Ideal for excitation of nucleic acids with UV of blue light. Unlike EtBR, SmartGlow is non mutagenic and does not require special methods of disposal. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4723 Supplier Braintree Scientific Supplier No. E4500PS
SmartGlow™ Nucleic Acid Stain Ideal for excitation of nucleic acids with UV of blue light. Unlike EtBR, SmartGlow is non mutagenic and does not require special methods of disposal. **Sold for research purposes only and not for human use**