Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit - Purple
Catalog No. 502105493
Encompass_Preferred
PerkinElmer introduces the NexION 1000 solution-ready sample introduction technology. This SMARTintro Sample Introduction Purple Kit was engineered for the widest range of NexION 1000 ICP-MS analyses and allows for quick and efficient changing of sample introduction hardware. This kit includes: MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Glass Nebulizer Type C (N8152373) Matrix Gas Line 9 in. (N8152374) Plasma Light Shield (N8152377) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Port (N8152389) Spray Chamber Light Shield (N8152425) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) Internal Standard Probe Qty. 2 (N8152452) PeriPump Union Male Fitting (N8152453) Spray Chamber Drain Line 1/4 28 Fitting (N8152456) One Piece 2.0 mm Quartz Torch (N8152472) Cassette Only Purple (N8152533)
Catalog No. 50-210-5493 Supplier Perkin Elmer US LLC Supplier No. N8150038
Add to Cart
Add to Cart
Request a Quote
PerkinElmer introduces the NexION 1000 solution-ready sample introduction technology. This SMARTintro Sample Introduction Purple Kit was engineered for the widest range of NexION 1000 ICP-MS analyses and allows for quick and efficient changing of sample introduction hardware. This kit includes: MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Glass Nebulizer Type C (N8152373) Matrix Gas Line 9 in. (N8152374) Plasma Light Shield (N8152377) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Port (N8152389) Spray Chamber Light Shield (N8152425) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) Internal Standard Probe Qty. 2 (N8152452) PeriPump Union Male Fitting (N8152453) Spray Chamber Drain Line 1/4 28 Fitting (N8152456) One Piece 2.0 mm Quartz Torch (N8152472) Cassette Only Purple (N8152533)