Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit Core Configuration - Green
Catalog No. 502111330
Encompass_Preferred
PerkinElmer introduces the NexION 2000 ICP-MS high matrix solution-ready sample introduction technology. Engineered for the analysis of environmental samples, this system utilizes a high sensitivity glass spray chamber and nebulizer, Peltier cooler, and large diameter injector to enhance the analysis of environmental samples with varying matrices. This kit includes: Glass Concentric Type C Nebulizer (N8152373) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Gas Port (N8152389) One-Piece 2.5 mm I.D. Injector Quartz Torch (N8152473) MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Matrix Gas Line 9 (N8152374) Plasma Light Shield (N8152377) PC3X Peltier Cyclonic Spray Chamber System (N8152382) MEINHARD Internal Standard Nebulizer Tee (N8152386) Spray Chamber Light Shield (N8152425), Green Torch Cassette (N8152431) Torch Injector Nut Removal Tool (N8152443) Internal Standard Probe(N8152452) PeriPump Union Male Fitting (N8152453) 0.8 mm I.D. PT
Catalog No. 50-211-1330 Supplier Perkin Elmer US LLC Supplier No. N8150035
Add to Cart
Add to Cart
Request a Quote
PerkinElmer introduces the NexION 2000 ICP-MS high matrix solution-ready sample introduction technology. Engineered for the analysis of environmental samples, this system utilizes a high sensitivity glass spray chamber and nebulizer, Peltier cooler, and large diameter injector to enhance the analysis of environmental samples with varying matrices. This kit includes: Glass Concentric Type C Nebulizer (N8152373) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Gas Port (N8152389) One-Piece 2.5 mm I.D. Injector Quartz Torch (N8152473) MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Matrix Gas Line 9 (N8152374) Plasma Light Shield (N8152377) PC3X Peltier Cyclonic Spray Chamber System (N8152382) MEINHARD Internal Standard Nebulizer Tee (N8152386) Spray Chamber Light Shield (N8152425), Green Torch Cassette (N8152431) Torch Injector Nut Removal Tool (N8152443) Internal Standard Probe(N8152452) PeriPump Union Male Fitting (N8152453) 0.8 mm I.D. PT