Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit Base Configuration - Blue
Catalog No. 502111323
Encompass_Preferred
PerkinElmer introduces the NexION 2000 ICP-MS Standard solution-ready sample introduction technology. It is engineered for the widest range of ICP-MS analyses. This kit includes: Glass Concentric Type C Nebulizer (N8152373) Glass Cyclonic C3 High Sensitivity Spray Chamber (N8152375) One-Piece 2.0 mm I.D. Injector Quartz Torch (N8152472) MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Plasma Light Shield (N8152377) MEINHARD Internal Standard Nebulizer Tee (N8152386) Spray Chamber Light Shield (N8152425) Blue Torch Cassette (N8152430) Torch Injector Nut Removal Tool (N8152443) Internal Standard Probe, Qty. 2 (N8152452) PeriPump Union Male Fitting (N8152453) 0.8 mm I.D. PTFE Drain Line Tubing with 1/4-28 Fitting (N8152456) Torch Injector Nut (N8152451)
Catalog No. 50-211-1323 Supplier Perkin Elmer US LLC Supplier No. N8150034
Add to Cart
Add to Cart
Request a Quote
PerkinElmer introduces the NexION 2000 ICP-MS Standard solution-ready sample introduction technology. It is engineered for the widest range of ICP-MS analyses. This kit includes: Glass Concentric Type C Nebulizer (N8152373) Glass Cyclonic C3 High Sensitivity Spray Chamber (N8152375) One-Piece 2.0 mm I.D. Injector Quartz Torch (N8152472) MEINHARD Nebulizer Sample Line (N8145503) PeriPump Tubing Starter Kit (N8150060) Plasma Light Shield (N8152377) MEINHARD Internal Standard Nebulizer Tee (N8152386) Spray Chamber Light Shield (N8152425) Blue Torch Cassette (N8152430) Torch Injector Nut Removal Tool (N8152443) Internal Standard Probe, Qty. 2 (N8152452) PeriPump Union Male Fitting (N8152453) 0.8 mm I.D. PTFE Drain Line Tubing with 1/4-28 Fitting (N8152456) Torch Injector Nut (N8152451)