Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit Semi-Conductor Configuration - White
Catalog No. 502111623
Encompass_Preferred
PerkinElmer introduces the2000 ICP-MS high purity solution-ready sample introduction technology. Engineered for analysis in the semiconductor industry, this high-performance system utilizes SilQ ultra high purity quartz spray chamber, torch, and injector to minimize contamination and delivers absolute accuracy and precision. This kit includes: Type C PFA ST3 Nebulizer (N8152378) SilQ Cyclonic C3 High Sensitivity Spray Chamber (N8152424) One-Piece 2.0 mm I.D. Injector SilQ Torch (N8152428) Female CTFE Barb Fitting (N8145018) PeriPump Starter Tubing Kit (N8150060) Torch Light Shield Disk (N8152377) PC3X Peltier Cyclonic Spray Chamber System (N8152382) PFA-ST Internal Standard Addition Tee (N8152423) Cyclonic Spray Chamber/Cassette UV Light Shield (N8152425) White Torch Cassette (N8152432) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) Internal Standard Probe, Qty. 2 (N8152452) 0.5 mm I.D. Sample Introduction Line for PFA-ST Nebulizer (N8152455) 0.8 mm I.D. PTF
Catalog No. 50-211-1623 Supplier Perkin Elmer US LLC Supplier No. N8150037
Add to Cart
Add to Cart
Request a Quote
PerkinElmer introduces the2000 ICP-MS high purity solution-ready sample introduction technology. Engineered for analysis in the semiconductor industry, this high-performance system utilizes SilQ ultra high purity quartz spray chamber, torch, and injector to minimize contamination and delivers absolute accuracy and precision. This kit includes: Type C PFA ST3 Nebulizer (N8152378) SilQ Cyclonic C3 High Sensitivity Spray Chamber (N8152424) One-Piece 2.0 mm I.D. Injector SilQ Torch (N8152428) Female CTFE Barb Fitting (N8145018) PeriPump Starter Tubing Kit (N8150060) Torch Light Shield Disk (N8152377) PC3X Peltier Cyclonic Spray Chamber System (N8152382) PFA-ST Internal Standard Addition Tee (N8152423) Cyclonic Spray Chamber/Cassette UV Light Shield (N8152425) White Torch Cassette (N8152432) Torch Injector Nut Removal Tool (N8152443) Torch Injector Nut (N8152451) Internal Standard Probe, Qty. 2 (N8152452) 0.5 mm I.D. Sample Introduction Line for PFA-ST Nebulizer (N8152455) 0.8 mm I.D. PTF