Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Kit Productivity Configuration - Black
Catalog No. 502111325
Encompass_Preferred
PerkinElmer introduces the NexION 2000 ICP-MS high throughput solution-ready sample introduction technology. Engineered with FAST capabilities to maximize ICP-MS productivity, this high-performance system utilizes superior sample introduction components and Peltier chiller to maximize accuracy and precision. This kit includes: 2.0 mm I.D. Demountable Quartz Injector (N8152376) Type C PFA ST3 Nebulizer (N8152378) Demountable Quartz Torch (N8152379) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Gas Port (N8152389) PC3X Peltier Cyclonic Spray Chamber System (N8152382) Torch Light Shield Disk (N8152377) Furrule-Nut Set with 1/4-28 Hi Flow Valve for 1/16 O.D. Tubing, Qty. 10 (N0777283) Ferrule-Nut Set with 1/4-28 Hi Flow Valves for 1/8 Tubing, Qty. 10 (N0777297) Sample Loop for Ports 1 and 4, 0.5 mL (N0777289) 0.8 mm I.D. Sample Loop, 300 µL (N0777430) 1.0 mm I.D. Sample Loop, 2 mL (N0777479) 1.0 mm I.D. Sample Loop, 4 mL (N0777480) 1.0 mm I.D. Sample Loop, 1 mL (N0777664)
Catalog No. 50-211-1325 Supplier Perkin Elmer US LLC Supplier No. N8150036
Add to Cart
Add to Cart
Request a Quote
PerkinElmer introduces the NexION 2000 ICP-MS high throughput solution-ready sample introduction technology. Engineered with FAST capabilities to maximize ICP-MS productivity, this high-performance system utilizes superior sample introduction components and Peltier chiller to maximize accuracy and precision. This kit includes: 2.0 mm I.D. Demountable Quartz Injector (N8152376) Type C PFA ST3 Nebulizer (N8152378) Demountable Quartz Torch (N8152379) Glass Cyclonic C3 High Sensitivity Spray Chamber with Matrix Gas Port (N8152389) PC3X Peltier Cyclonic Spray Chamber System (N8152382) Torch Light Shield Disk (N8152377) Furrule-Nut Set with 1/4-28 Hi Flow Valve for 1/16 O.D. Tubing, Qty. 10 (N0777283) Ferrule-Nut Set with 1/4-28 Hi Flow Valves for 1/8 Tubing, Qty. 10 (N0777297) Sample Loop for Ports 1 and 4, 0.5 mL (N0777289) 0.8 mm I.D. Sample Loop, 300 µL (N0777430) 1.0 mm I.D. Sample Loop, 2 mL (N0777479) 1.0 mm I.D. Sample Loop, 4 mL (N0777480) 1.0 mm I.D. Sample Loop, 1 mL (N0777664)