Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Module, Black, with Demountable 2.0 mm I.D. Quartz Injector and Quartz Torch
Catalog No. 502112034
Encompass_Preferred
This module, incorporated in the NexION 2000 ICP-MS Productivity Configuration Kit, includes: Cassette Piece (Black) (N8152433) 2 mm I.D. Demountable Quartz Injector (N8152376) Demontable Quartz Torch (N8152379) Torch Injector Nut (N8152451) PTFE Torch Light Shield (N8152377)
Catalog No. 50-211-2034 Supplier Perkin Elmer US LLC Supplier No. N8152440
This module, incorporated in the NexION 2000 ICP-MS Productivity Configuration Kit, includes: Cassette Piece (Black) (N8152433) 2 mm I.D. Demountable Quartz Injector (N8152376) Demontable Quartz Torch (N8152379) Torch Injector Nut (N8152451) PTFE Torch Light Shield (N8152377)