Learn More
Research Products International Corp SmartPicks, Standard, Semi Point, 2.5 inches long, 1600 per case
SDP
Catalog No. 501978023
Encompass
smartPicks Wooden Inoculation are manufactured of naturally biodegradable 100% USA grown white birch, autoclavable and naturally biodegradable. Available in both a semi-pointed and needle pointed tip for various applications. Short 1.28" tips are ideal for use with 1.5 ml micro-tubes.
Catalog No. 50-197-8023 Supplier Research Products International Corp Supplier No. 249916
smartPicks Wooden Inoculation are manufactured of naturally biodegradable 100% USA grown white birch, autoclavable and naturally biodegradable. Available in both a semi-pointed and needle pointed tip for various applications. Short 1.28" tips are ideal for use with 1.5 ml micro-tubes.