Learn More
Instrotek Inc SMARTRACKER
SDP
Catalog No. NC3662199
Encompass
  • Hamburg wheel tracker designed for asphalt mixture evaluation
  • Allows simultaneous wet and dry testing
  • Features a retractable wheel system
  • Independent motor control for wheel drive
  • On-board Windows touchscreen computer for easy operation
  • Data download options via USB, SD, or network connectivity
  • Meets AASHTO T-324 and NCHRP 20-07 Project 361 standards
  • Constructed with rigid steel frame and stainless steel components
  • Weighs 658 kg (1450 lbs)
  • Dimensions: 1.27 m height, 1.37 m width x 1.22 m depth
Catalog No. NC3662199 Supplier Instrotek Inc Supplier No. 294.3038
Add to Cart
Add to Cart
Request a Quote
  • Hamburg wheel tracker designed for asphalt mixture evaluation
  • Allows simultaneous wet and dry testing
  • Features a retractable wheel system
  • Independent motor control for wheel drive
  • On-board Windows touchscreen computer for easy operation
  • Data download options via USB, SD, or network connectivity
  • Meets AASHTO T-324 and NCHRP 20-07 Project 361 standards
  • Constructed with rigid steel frame and stainless steel components
  • Weighs 658 kg (1450 lbs)
  • Dimensions: 1.27 m height, 1.37 m width x 1.22 m depth