Learn More
Alkali Scientific SmartReader Thermal Printer
SDP
Catalog No. 502388847
Encompass_Preferred
The Accuris SmartReader 96 is a welcome addition to any lab that is routinely measuring concentration or absorbance in 96 well plates. The instrument’s 7 inch touch screen, intuitive software and graphical interface makes it easy to use as a standalone instrument (a separate computer and additional software is not required).
Catalog No. 50-238-8847 Supplier Alkali Scientific Supplier No. W9600TP
The Accuris SmartReader 96 is a welcome addition to any lab that is routinely measuring concentration or absorbance in 96 well plates. The instrument’s 7 inch touch screen, intuitive software and graphical interface makes it easy to use as a standalone instrument (a separate computer and additional software is not required).