Learn More
Pike Technologies SmartSeal Transmission Cell, 0.05 mm pathlength, BaF2 windows, 0.025 mL volume, 13 mm clear aperture, 2" x 3" slide mount, compatible with FTIR spectrometers, Luer-Lok™ fittings
SDP
Catalog No. NC3364378
Encompass_Preferred
SmartSeal Transmission Cell, 0.05 mm pathlength, BaF2 windows, 0.025 mL volume, 13 mm clear aperture, 2" x 3" slide mount, compatible with FTIR spectrometers, Luer-Lok™ fittings
Catalog No. NC3364378 Supplier Pike Technologies Supplier No. 1654002
SmartSeal Transmission Cell, 0.05 mm pathlength, BaF2 windows, 0.025 mL volume, 13 mm clear aperture, 2" x 3" slide mount, compatible with FTIR spectrometers, Luer-Lok™ fittings