Learn More
Research Products International Corp Disposable smartSpatula, 210mm Long, Natural, 300 per Package
SDP
Catalog No. 50476568
Encompass
This disposable SmartSpatula provides the most innovative design for a new multi-purpose lab tool. Features a hollow core handle with both a scoop and spatula end for convenient transfer of collection of materials. Can also be used to stir solutions. Flexible ends become shatter resistant after freezing in liquid nitrogen and readly form to an object to scrape or collect samples. Manufactured from chemical resistant polypropylene and can be autoclaved. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc
Catalog No. 50-476-568 Supplier Research Products International Corp Supplier No. 249901
This disposable SmartSpatula provides the most innovative design for a new multi-purpose lab tool. Features a hollow core handle with both a scoop and spatula end for convenient transfer of collection of materials. Can also be used to stir solutions. Flexible ends become shatter resistant after freezing in liquid nitrogen and readly form to an object to scrape or collect samples. Manufactured from chemical resistant polypropylene and can be autoclaved. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc