Learn More
Alkali Scientific Alkali Scientific SmartWasher™ 96 Microplate Washer (MW9600)
SDP
Catalog No. 503029517
Encompass_Preferred
The Alkali Scientific SmartWasher™ 96 Microplate Washer (MW9600) automates the process of ELISA plate washing so assays can be prepared quickly, accurately, and with confidence for absorbance reading.  It’s the perfect companion to our popular SmartReader™ 96 microplate reader. Precision control of aspiration needle position– Accurate to 0.1mm– Minimal residual liquid, ≤1μl/well Auto rinse function eliminates cross contamination Up to 100 wash protocols can be customized and stored Liquid level sensors in wash and waste bottles Wash protocols can be transferred via flash drive
Catalog No. 50-302-9517 Supplier Alkali Scientific Supplier No. RS9600
The Alkali Scientific SmartWasher™ 96 Microplate Washer (MW9600) automates the process of ELISA plate washing so assays can be prepared quickly, accurately, and with confidence for absorbance reading.  It’s the perfect companion to our popular SmartReader™ 96 microplate reader. Precision control of aspiration needle position– Accurate to 0.1mm– Minimal residual liquid, ≤1μl/well Auto rinse function eliminates cross contamination Up to 100 wash protocols can be customized and stored Liquid level sensors in wash and waste bottles Wash protocols can be transferred via flash drive