Learn More
Kyfora Bio Smashgene Transfection Reagent 10mL bottle transfection reagent 1/EA
Supplier:  Kyfora Bio 1457110
Encompass_Preferred
SmashGene is a next-generation transfection reagent engineered to simplify workflows while delivering stronger performance and smarter economics. Designed to eliminate the need for enhancers, it enables a more streamlined and cost-effective transfection process without sacrificing yield. By reducing complexity and minimizing variables, SmashGene accelerates development timelines and improves reproducibility from early research through scale-up. It consistently produces higher titers than leading competitor reagents while requiring fewer components, resulting in lower overall production costs through reduced bioreactor runs and simplified downstream processing. Compatible with both adherent and suspension cell systems across a wide range of media conditions, SmashGene offers exceptional flexibility for diverse process strategies. Fully synthetic and manufactured in the USA, it ensures reliable supply, consistent quality, and greater control from flask to bioreactor.
Catalog No. 50-005-74089