Learn More
Lonza Walkersville SmBM Smooth Muscle Cell Growth Basal Medium, 500 mL
Catalog No. NC9586602
Encompass
SmBM -2 is a Smooth Muscle Cell basal medium sold separately. This can be used with 1 kit of CC-4149 (smooth muscle cell growth medium SingleQuots Supplements and Growth Factors). The full SmGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3182. SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Clonetic Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x Bottle SmBM Basal Medium, 500 mL
Catalog No. NC9586602 Supplier Lonza Walkersville Supplier No. CC3181
SmBM -2 is a Smooth Muscle Cell basal medium sold separately. This can be used with 1 kit of CC-4149 (smooth muscle cell growth medium SingleQuots Supplements and Growth Factors). The full SmGM -2 BulletKit consisting of basal medium and supplements can be purchased together using the catalog number CC-3182. SmGM-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more. This kit is also utilized to support the maintenance of intestinal cells. SmGM-2 medium contains 5% FBS.Clonetic Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.Content:1 x Bottle SmBM Basal Medium, 500 mL