Learn More
McMaster-Carr SMC Compressed Air Regulators
Catalog No. NC2385648
Encompass
SMC compressed air regulators offer precise pressure control for industrial pneumatic applications. With +/-0.5% F.S. accuracy and up to 1700 L/min flow, these compact regulators come in 1/8" to 1" sizes, handle up to 16 bar inlet pressure, and offer 0-1 MPa outlet ranges. Features include knob adjustment, gauge options, and modular accessories.
Catalog No. NC2385648 Supplier McMaster-Carr Supplier No. 4989K501
SMC compressed air regulators offer precise pressure control for industrial pneumatic applications. With +/-0.5% F.S. accuracy and up to 1700 L/min flow, these compact regulators come in 1/8" to 1" sizes, handle up to 16 bar inlet pressure, and offer 0-1 MPa outlet ranges. Features include knob adjustment, gauge options, and modular accessories.