Learn More
Aviva Systems Biology Corp SMC1A Antibody - C-terminal region (ARP38780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMC1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SMC1A. Alternate names: CDLS2, DKFZp686L19178, DXS423E, KIAA0178, MGC138332, SB1.8, SMC1, SMC1L1, SMC1alpha, SMCBApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56795