Learn More
Aviva Systems Biology Corp SMC4 Antibody - N-terminal region (ARP46130_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46130_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMC4. Alternate names: CAPC, SMC4L1, hCAP-C, CAP-C, SMC-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59241