Learn More
ABclonal Technology SMC4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A139331000UL
Encompass_Preferred
This gene belongs to the structural maintenance of chromosomes (SMC) gene family. Members of this gene family play a role in two changes in chromosome structure during mitotic segregation of chromosomes- chromosome condensation and sister chromatid cohesion. The protein encoded by this gene is likely a subunit of the 13S condensin complex, which is involved in chromosome condensation. A pseudogene related to this gene is located on chromosome 2.
Catalog No. 50-001-06793