Learn More
Aviva Systems Biology Corp SMC6 Antibody - C-terminal region (ARP64076_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64076_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMC6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human SMC6. Alternate names: FLJ22116, FLJ35534, SMC6L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-288-165