Learn More
TARGETMOL CHEMICALS INC SMCC-DM1 5MG
SDP
Catalog No. 502879386
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. SMCC-DM1 is a drug-coupler coupler consisting of a potent microtubule disrupter, DM1, and a coupler, SMCC, for the preparation of antibody-drug couplers. Purity 99.25%
Catalog No. 50-287-9386 Supplier TARGETMOL CHEMICALS INC Supplier No. T168995MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. SMCC-DM1 is a drug-coupler coupler consisting of a potent microtubule disrupter, DM1, and a coupler, SMCC, for the preparation of antibody-drug couplers. Purity 99.25%