Learn More
Aviva Systems Biology Corp MIEF1 Antibody - N-terminal region (ARP49422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMCR7L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMCR7L. Alternate names: FLJ20232, HSU79252, dJ1104E15.3, MID51, MIEF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60081