Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human cTNi High Sens
Catalog No. 502906217
Encompass
The SMC(TM) human cTnI High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences 1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification. 2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.
Catalog No. 50-290-6217 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03015400
The SMC(TM) human cTnI High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences 1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification. 2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.