Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human IL 13 High Sen
Catalog No. 502906209
Encompass
The SMC(TM) Human IL-13 High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences 1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification. 2) The Erenna and SMCxPRO immunoassay systems detect analytes using Single Molecule Counting (SMC) technology.
Catalog No. 50-290-6209 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03015600
The SMC(TM) Human IL-13 High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences 1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification. 2) The Erenna and SMCxPRO immunoassay systems detect analytes using Single Molecule Counting (SMC) technology.