Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human IL 17A High Se
Catalog No. 502905204
Encompass
The SMC(TM) human IL-17A High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.Assay protocols are similar to existing sandwich ELISA methods with two key differences1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification.2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.
Catalog No. 50-290-5204 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03015900
The SMC(TM) human IL-17A High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.Assay protocols are similar to existing sandwich ELISA methods with two key differences1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification.2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.