Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human Interleukin 6
Catalog No. 502905468
Encompass
Interleukin 6 (IL-6) is secreted by T cells and macrophages and acts as both a pro-inflammatory cytokine and an anti-inflammatory myokine. Some of its functions include the stimulation of the immune response during infection induction of fever in autoimmune infectious and non-infectious diseases the stimulation of acute phase reactions differentiation of T cells proliferation and differentiation of B-cells the development of blood cells as well as the activation of osteoclasts and osteoporosis and the secretion of vascular endothelium growth factor (VEfG) to increase blood vessel growth and vascular permeability in inflammation. It is important for a short-term defense against infection injury and inflammation however dysregulation of IL-6 results in disease.IL-6 is important in the study of diseases such as diabetes lupus multiple myeloma rheumatoid arthritis and some cancers such as prostate and pancreatic cancer. Conversely deficiency of IL-6 can have an effect on immune
Catalog No. 50-290-5468 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03015500
Interleukin 6 (IL-6) is secreted by T cells and macrophages and acts as both a pro-inflammatory cytokine and an anti-inflammatory myokine. Some of its functions include the stimulation of the immune response during infection induction of fever in autoimmune infectious and non-infectious diseases the stimulation of acute phase reactions differentiation of T cells proliferation and differentiation of B-cells the development of blood cells as well as the activation of osteoclasts and osteoporosis and the secretion of vascular endothelium growth factor (VEfG) to increase blood vessel growth and vascular permeability in inflammation. It is important for a short-term defense against infection injury and inflammation however dysregulation of IL-6 results in disease.IL-6 is important in the study of diseases such as diabetes lupus multiple myeloma rheumatoid arthritis and some cancers such as prostate and pancreatic cancer. Conversely deficiency of IL-6 can have an effect on immune