Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Human TNfalpha High
Catalog No. 502905236
Encompass
The SMC(TM) human TNfalpha High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.Assay protocols are similar to existing sandwich ELISA methods with two key differences1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification.2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.
Catalog No. 50-290-5236 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03016300
The SMC(TM) human TNfalpha High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format.Assay protocols are similar to existing sandwich ELISA methods with two key differences1) Elution buffer disrupts the sandwich separating the labeled detection antibody for quantification.2)The Erenna and SMCxPRO(TM) immunoassay systems detect analytes using Single Molecule Counting (SMC(TM)) technology.