Learn More
Sigma Aldrich Fine Chemicals Biosciences SMCTM Plate Based Immunoas
Catalog No. 502905469
Encompass
Take advantage of Single Molecule Counting (SMC(TM)) technology while developing your own application-specific immunoassays in plate format. This kit provides everything you need to build your own high performing SMC(TM) immunoassay. It includes buffers reagents and optimized protocol for plate assay development.
Catalog No. 50-290-5469 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03012800
Take advantage of Single Molecule Counting (SMC(TM)) technology while developing your own application-specific immunoassays in plate format. This kit provides everything you need to build your own high performing SMC(TM) immunoassay. It includes buffers reagents and optimized protocol for plate assay development.