Learn More
Medchemexpress LLC Smcy HY peptide (738-746) | 207113-06-2 | 1167.34 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP18995MG
Encompass_Preferred
Smcy HY peptide (738-746) is an H2-Dᵇ-restricted peptide corresponding to amino acids 738-746 of the Smcy protein, targeting MHC in the immunology/inflammation pathway. It has a sequence of Lys-Cys-Ser-Arg-Asn-Arg-Gln-Tyr-Leu.
  • H2-Dᵇ-restricted peptide
  • Corresponds to amino acids 738-746 of Smcy protein
  • Targets MHC
  • Involved in immunology/inflammation pathway
  • Can be stored as powder for up to 2 years at -80°C
  • Can be stored in solvent for up to 6 months at -80°C
  • Recommended storage conditions are sealed, away from moisture and light, under nitrogen
Catalog No. 50-004-23146