Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Smcy HY Peptide (738-746) | 207113-06-2 | 98.2% | 1167.34 | 10 MG
SDP

Catalog No. 5000424169
Encompass_Preferred

Smcy HY Peptide (738-746) is a H2-Db-restricted peptide corresponding to amino acids 738-746 of Smcy protein. This product is for research use only.

  • Purity: 98.2%
  • Molecular weight: 1167.34
  • Sequence: Lys-Cys-Ser-Arg-Asn-Arg-Gln-Tyr-Leu
  • Soluble in H2O at 100 mg/mL (85.66 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light, under nitrogen)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light, under nitrogen)

Catalog No. 50-004-24169 Supplier Medchemexpress LLC Supplier No. HYP189910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Smcy HY Peptide (738-746) is a H2-Db-restricted peptide corresponding to amino acids 738-746 of Smcy protein. This product is for research use only.

  • Purity: 98.2%
  • Molecular weight: 1167.34
  • Sequence: Lys-Cys-Ser-Arg-Asn-Arg-Gln-Tyr-Leu
  • Soluble in H2O at 100 mg/mL (85.66 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light, under nitrogen)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light, under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.