Learn More
Medchemexpress LLC SMD-3040 (formate) | 3033109-92-8 | 99.8% | 989.2 g/mol | C53H68N10O7S | 10 MG
SDP
Catalog No. 5000288092
Encompass_Preferred
SMD-3040 formate is a research-grade PROTAC degrader that selectively targets SMARCA2, producing robust degradation (DC50: 12 nM; Dmax: 91%) and inhibiting proliferation of certain tumor cell lines. It has demonstrated in vivo antitumor activity in xenograft models and is supplied as the formate salt for laboratory research (not for human use).
  • Potent SMARCA2 degradation (DC50: 12 nM; Dmax: 91%).
  • Demonstrated in vivo antitumor activity in xenograft models.
  • High chemical purity (~99.8%).
  • Supplied as the formate salt for laboratory use.
  • Suitable for cell-based and in vivo pharmacology studies (research use only).
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-88092 Supplier Medchemexpress LLC Supplier No. HY156568B10MG
SMD-3040 formate is a research-grade PROTAC degrader that selectively targets SMARCA2, producing robust degradation (DC50: 12 nM; Dmax: 91%) and inhibiting proliferation of certain tumor cell lines. It has demonstrated in vivo antitumor activity in xenograft models and is supplied as the formate salt for laboratory research (not for human use).
  • Potent SMARCA2 degradation (DC50: 12 nM; Dmax: 91%).
  • Demonstrated in vivo antitumor activity in xenograft models.
  • High chemical purity (~99.8%).
  • Supplied as the formate salt for laboratory use.
  • Suitable for cell-based and in vivo pharmacology studies (research use only).
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).