Learn More
TARGETMOL CHEMICALS INC SMER28 25MG
SDP
Catalog No. 502576989
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SMER28 is a small-molecule enhancer (SMER) of autophagy inducing autophagy independently of rapamycin in mammalian cells via an mTOR-independent mechanism and prevented the accumulation of amyloid beta peptide. purity: 99%
Catalog No. 50-257-6989 Supplier TARGETMOL CHEMICALS INC Supplier No. T315525MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SMER28 is a small-molecule enhancer (SMER) of autophagy inducing autophagy independently of rapamycin in mammalian cells via an mTOR-independent mechanism and prevented the accumulation of amyloid beta peptide. purity: 99%