Learn More
Medchemexpress LLC HY-10949 5mg , SMER3 CAS:67200-34-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109495MG
Encompass_Preferred
HY-10949 5mg SMER3 CAS: 67200-34-4 SMER3, a Rapamycin enhancer, is a selective Skp1-Cullin-F-box (SCF)Met30 ubiquitin ligase inhibitor. SMER3 enhances Rapamycin's growth inhibitory effect by inhibition of SCFMet30. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1284