Learn More
Abcam SMER3, SCFMet30 ubiquitin E3 ligase inhibitor, 10MG
Catalog No. 502705589
Encompass_Preferred
MW 224.17 Da, Purity >98%. Cell permeable, selective SCFMet30 ubiquitin E3 ligase inhibitor. Enhances the action of rapamycin. Induces cell cycles arrest (IC50 = 10-30 μM). Active *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5589 Supplier Abcam Supplier No. AB14459610MG
MW 224.17 Da, Purity >98%. Cell permeable, selective SCFMet30 ubiquitin E3 ligase inhibitor. Enhances the action of rapamycin. Induces cell cycles arrest (IC50 = 10-30 μM). Active *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement