Learn More
Cayman Chemical S-Methyl-L-Cysteine-S-oxide, Purity: ≥98%, CAS: 6853-87-8, 100 mg
SDP
Catalog No. 502967704
Encompass_Preferred
An analog of alliin found in some cruciferous vegetables that can be converted through alliinase activity into a volatile thiosulfinate
Catalog No. 50-296-7704 Supplier Cayman Chemical Supplier No. 17600100 MG
An analog of alliin found in some cruciferous vegetables that can be converted through alliinase activity into a volatile thiosulfinate