Learn More
Cayman Chemical S-methyl-L-Thiocitrulline (hydrochloride), Purity: ≥97%, CAS: 209589-59-3, 100 mg
SDP
Catalog No. 502894837
Encompass_Preferred
S-methyl-L-Thiocitrulline is a potent NOS inhibitor with selectivity toward the neuronal isoform compared to eNOS and iNOS. For human enzymes, it exhibits Ki values of 1.2, 11, and 40 nM for nNOS, eNOS, and iNOS, respectively.{1325,1293}
Catalog No. 50-289-4837 Supplier Cayman Chemical Supplier No. 80585100 MG
S-methyl-L-Thiocitrulline is a potent NOS inhibitor with selectivity toward the neuronal isoform compared to eNOS and iNOS. For human enzymes, it exhibits Ki values of 1.2, 11, and 40 nM for nNOS, eNOS, and iNOS, respectively.{1325,1293}