Learn More
ABclonal Technology SMG1 Rabbit mAb
SDP
Catalog No. 5000008236
Encompass_Preferred
This gene encodes a protein involved in nonsense-mediated mRNA decay (NMD) as part of the mRNA surveillance complex The protein has kinase activity and is thought to function in NMD by phosphorylating the regulator of nonsense transcripts 1 protein Alternatively spliced transcript variants have been described but their full-length nature has yet to be determined
Catalog No. 50-000-08236 Supplier ABclonal Technology Supplier No. A19253100UL
This gene encodes a protein involved in nonsense-mediated mRNA decay (NMD) as part of the mRNA surveillance complex The protein has kinase activity and is thought to function in NMD by phosphorylating the regulator of nonsense transcripts 1 protein Alternatively spliced transcript variants have been described but their full-length nature has yet to be determined